Få mere at vide
Furaltadone 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Furaltadone |
| CAS | 139-91-3 |
| Analyt- eller komponentnavne | Furaltadone |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C13 H16 N4 O6 |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | 2-Oxazolidinone, 5-(4-morpholinylmethyl)-3-[[(5-nitro-2-furanyl)methylene]amino]-, 2-Oxazolidinone, 5-(morpholinomethyl)-3-[(5-nitrofurfurylidene)amino]- (6CI,7CI,8CI), 3-(5-Nitro-2-furfurylideneamino)-5-(4-morpholinomethyl)-2-oxazolidone, 5-(4-Morpholinomethyl)-3-(5-nitro-2-furfurylideneamino)-2-oxazolidinone, 5-(N-Morpholinomethyl)-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylidenamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidinone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)-2-oxazolidone, 5-Morpholinomethyl-3-(5-nitrofurfurylideneamino)oxazolidone, Altabactina, Altafur, Anauran, F 150, Fur-Novo, Furaltadon, Furaltadone, Furantoina, Furazolin, Furazoline, Furitale, Furlate, Furlidon, Furmethanol, Furmethonol, Furmetonol, Furmetox, Ibifur, Medifuran, N-(5-Nitro-2-furylidene)-3-amino-5-(N'-morpholinomethyl)-2-oxazolidone, NF 260, NSC 42388, Nitraldone, Nitrofurmethone, Nitrofurmeton, Otifuril, Sepsinol, Ultrafur, Unifur, Valsyn |
| InChI formel | InChI=1S/C13H16N4O6/c18-13-16(14-7-10-1-2-12(22-10)17(19)20)9-11(23-13)8-15-3-5-21-6-4-15/h1-2,7,11H,3-6,8-9H2 |
| SMIL | [O-][N+](=O)c1oc(C=NN2CC(CN3CCOCC3)OC2=O)cc1 |
| Vis mere |