Learn More
SCAT™ Funnels and Adapters for Barrels and Drums with Mechanical Level Control
Robust SCAT™ 'ARNOLD' Safety Funnels with adapters and mechanical level controls for use on various drums with different thread types. Also available in electrically dissipative variants for the safe disposal of explosive liquid waste.
1765.00 DKK - 3425.00 DKK
Specifications
For Use With (Equipment) SCAT ARNOLD Funnel
View More Specs
Products 4
Product Code Brand Includes Product Type Price Quantity & Availability  
Product Code Brand Includes Product Type Price Quantity & Availability  
17893510
View Documents
SCAT™
107889
Adapter, Mechanical Level Control Funnel Adapter
2295.00 DKK
Each
Only null left
Estimated Shipment: 14-07-2025
Add to basket
17838733
View Documents
SCAT™
317631
Lid, Splash Guard, Removable Sieve, Grounding Cable Safety Funnel
1765.00 DKK
Each
Only null left
Estimated Shipment: 10-07-2025
Add to basket
17848743
View Documents
SCAT™
317650
Lid, Splash Guard, Removable Sieve, Grounding Cable, Adapter, Mechanical Level Control Safety Funnel
3425.00 DKK
Each
Only null left
Estimated Shipment: 10-07-2025
Add to basket
17868743
View Documents
SCAT™
317660
Lid, Splash Guard, 2 Removable Sieves, Adapter, Mechanical Level Control Safety Funnel
3065.00 DKK
Each
Only null left
Estimated Shipment: 10-07-2025
Add to basket
Description
Description
  • Safety funnels based on the SCAT™ 'ARNOLD' funnel series
  • Ready-to-use sets consisting of safety funnels and barrel adapters
  • Made of high-quality plastics for durable and long-lasting use
  • Adapters can be combined with all GL45 safety funnels of the 'ARNOLD' and 'MARCO' series
  • Available for all common thread types and sizes
  • Also available in electrically conductive material for hazardous areas
Specifications
Specifications
SCAT ARNOLD Funnel
Documents
Product Certifications
Product Content Correction
Product Title
SCAT™ Funnels and Adapters for Barrels and Drums with Mechanical Level Control