Få mere at vide
Fumaric Acid Monomethyl Ester, TRC
Artikelnummer. 30138434
Skift visning
Mængde:
50 mg
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138434 50 mg
30135495 25 g
30004194 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138434 Leverandør Toronto Research Chemicals Leverandørnr. TRCF50039050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2756-87-8
Molekylær formel C5 H6 O4
Molekylvægt (g/mol) 130.1
Synonym Monomethyl Fumarate, (2E)-2-Butenedioic acid 1-methyl ester, (E)-2-Butenedioic acid monomethyl ester, Fumaric acid methyl ester, Fumaric acid monomethyl ester, Methyl hydrogen fumarate, Monomethyl fumarate
IUPAC navn (E)-4-methoxy-4-oxobut-2-enoic acid
SMIL COC(=O)\C=C\C(=O)O
Tekniske data
Kemisk navn eller materiale Monomethyl Fumarate
Mængde 50 mg
InChI formel InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+
Formel vægt 130.0266