missing translation for 'onlineSavingsMsg'
Få mere at vide

Fumaric Acid Monomethyl Ester-d3, TRC

Artikelnummer. 30405447
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405447 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405447 Leverandør Toronto Research Chemicals Leverandørnr. TRCF50039210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Fumaric Acid Monomethyl Ester-d3
CAS 1616345-41-5
Renhedsgrad noter HPLC
Molekylær formel C5H3D3O4
Mængde 10 mg
Synonym (2E)-2-Butenedioic Acid 1-Methyl Ester-d3, (E)-2-Butenedioic Acid Monomethyl Ester-d3, Fumaric Acid Methyl Ester-d3, Methyl Hydrogen Fumarate-d3, Monomethyl Fumarate-d3
InChI formel InChI=1S/C5H6O4/c1-9-5(8)3-2-4(6)7/h2-3H,1H3,(H,6,7)/b3-2+/i1D3
SMIL OC(/C=C/C(OC([2H])([2H])[2H])=O)=O
IUPAC navn (E)-4-(methoxy-d3)-4-oxobut-2-enoic acid
Molekylvægt (g/mol) 133.12
Formel vægt 133.05
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.