Få mere at vide
Fumaric Acid Monoethyl-d5 Ester, TRC
Artikelnummer. 30403936
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30403936 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403936 Leverandør Toronto Research Chemicals Leverandørnr. TRCF500387100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fumaric Acid Monoethyl-d5 Ester
CAS 1266398-64-4
Molekylær formel C6H3D5O4
Mængde 100 mg
Synonym (2E)-2-Butenedioic Acid 1-Ethyl-d5 Ester, (2E)-2-Butenedioic Acid Monoethyl-d5 Ester, Fumaric Acid-d5 Ethyl Ester, (E)-4-(Ethoxy-d5)-4-oxo-2-butenoic Acid, Ethyl-d5 Hydrogen Fumarate, Monoethyl-d5 Fumarate, Monoethyl-d5 Fumerate, Monoethyl-d5 trans-2-Butenedioate
InChI formel InChI=1S/C6H8O4/c1-2-10-6(9)4-3-5(7)8/h3-4H,2H2,1H3,(H,7,8)/i1D3,2D2
SMIL [2H]C([2H])([2H])C([2H])([2H])OC(=O)C=CC(=O)O
IUPAC navn 4-oxo-4-(1,1,2,2,2-pentadeuterioethoxy)but-2-enoic acid
Molekylvægt (g/mol) 149.16
Formel vægt 149.0736