Få mere at vide
Fumaric Acid-d2, TRC
Artikelnummer. 30408056
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30408056 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408056 Leverandør Toronto Research Chemicals Leverandørnr. TRCF5003841G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fumaric Acid-d2
CAS 24461-32-3
Molekylær formel C4 D2 H2 O4
Mængde 1 g
Synonym (2E)-2-Butenedioic Acid-d2, (2E)-But-2-enedioic Acid-d2, (E)-2-Butenedioic Acid-d2, 2-(E)-Butenedioic Acid-d2, (E)-Butenedioic Acid-d2, Allomaleic Acid-d2, Bakeshure 451-d2, Bakeshure 470-d2, Boletic Acid-d2, FC 33-d2, Lichenic Acid-d2, NSC 2752-d2, Petrom-d2, trans-1,2-Ethylenedicarboxylic Acid-d2, trans-2-Butenedioic Acid-d2, trans-Butenedioic Acid-d2
InChI formel InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+/i1D,2D
SMIL [2H]\C(=C(\[2H])/C(=O)O)\C(=O)O
IUPAC navn (E)-2,3-dideuteriobut-2-enedioic acid
Molekylvægt (g/mol) 118.08
Formel vægt 118.0235