Få mere at vide
3M™ Fuel™ Sikkerhedsbriller
Shop alle 3M produkter
Linsefarve:
Blå, Spejl
Bronze
Indoor/Outdoor, Mirror
Red, Mirror
Ramme farve:
Bronze
Sølv
Silver
Pakningsstørrelse:
Pakke med 20
Beskrivelse
- Safety glasses designed to be worn at work, at home or during outdoor recreational activities
- The frame's molded lens supports prevent the lenses from being dislodged by a strong frontal impact
- Ventilation system on nose bridge and temples to help prevent fogging
Tekniske data
Tekniske data
| Certificeringer/compliance | CE Approved, EN 166:2001 |
| Linsebelægning | Scratch-resistant |
| Oversigt | Red Mirror coating for increased durability and improved vision |
| Linsefarve | Red, Mirror |
| Produkttype | Safety glasses |
| Ramme farve | Bronze |
| Linse materiale | Polycarbonate |