missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Fucosyl GM1 Antibody (BMS-986012) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28503JF669
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Fucosyl GM1 Monoclonal antibody specifically detects Fucosyl GM1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Tekniske data
| Fucosyl GM1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Human | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Functional Assay | |
| BMS-986012 | |
| 50mM Sodium Borate | |
| Fucosyl GM1 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion