Learn More
R&D Systems™ FUBP3 293T Cell Transient Overexpression Lysate
Product Code. 18610011 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
Product Code. Quantity Unit Size
18610011 0.1 mL 0.1mL
1 options
Product Code. 18610011 Supplier R&D Systems™ Supplier No. H00008939T01
 Restricted Item
Forventet forsendelse: 07-12-2026
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 0.1 mL
Content And Storage Store at -80C. Avoid freeze-thaw cycles.
Product Line Novus
Target FUBP3
Synonym far upstream element (FUSE) binding protein 3, far upstream element-binding protein 3, FBP3, FLJ25229, FUSE-binding protein 3
Gene Symbol FUBP3