Få mere at vide
Tocris Bioscience™ FTY 720
Potent S1P receptor agonist; also immunosuppressant
Brand:  Tocris Bioscience™ 6176/50
Encompass
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Produktkode 15757246
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
FTY 720 is a S1P receptor agonist; undergoes phosphorylation by sphingosine kinase 2, which then binds S1P1,3,4 and 5 receptors (EC50 values are 0.3 - 0.6 nM for S1P1, 4 and 5 and ∽3 nM for S1P3). Immunosuppresant; sequesters lymphocytes in lymph nodes, reducing autoimmune response. Suppresses the development of EAE (an in vivo model of MS). Orally bioavailable and blood-brain barrier permeable.
Tekniske data
Tekniske data
Potent S1P Receptor Agonist, also Immunosuppressant
162359-56-0
343.93g/mol
Store at -20°C
50 mg
Sphingosine-1-phosphate Receptor
C19H33NO2.HCl
Fingolimod, Gilenya
Sphingosine-1-phosphate Receptor Agonists
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Produkttitel
Vælg et problem
For Research Use Only