Få mere at vide
Fructosyl Glycine Alpha/Beta Mixture (Mixture of Diastereomers), TRC
Artikelnummer. 17978764
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17978764 10 mg
17988494 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17978764 Leverandør Toronto Research Chemicals Leverandørnr. TRCF79575010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60644-20-4
Molekylær formel C8 H15 N O7
Molekylvægt (g/mol) 237.207
Synonym N-(1-Deoxy-β-D-fructopyranos-1-yl)glycine, 1-[(Carboxymethyl)amino]-1-deoxy-β-D-fructopyranose
IUPAC navn 2-[[(3S,4R,5R)-2,3,4,5-tetrahydroxytetrahydropyran-2-yl]methylamino]acetic acid
SMIL O[C@@H]1COC(O)(CNCC(=O)O)[C@@H](O)[C@@H]1O
Tekniske data
Kemisk navn eller materiale Fructosyl Glycine Alpha/Beta Mixture (Mixture of Diastereomers)
Renhedsgrad noter HPLC
InChI formel InChI=1S/C8H15NO7/c10-4-2-16-8(15,7(14)6(4)13)3-9-1-5(11)12/h4,6-7,9-10,13-15H,1-3H2,(H,11,12)/t4-,6-,7+,8?/m1/s1
Mængde 10 mg
Formel vægt 237.0849
Procent renhed >95%