Få mere at vide
Fructose-proline (mixture of diastereomers), TRC
Artikelnummer. 30006145
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006145 100 mg
30003258 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006145 Leverandør Toronto Research Chemicals Leverandørnr. TRCF792565100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 29118-61-4
Molekylær formel C11H19NO7
Molekylvægt (g/mol) 277.27
Synonym 1-(1-Deoxy-D-fructos-1-yl)-L-proline, (S)-1-(2-Carboxy-1-pyrrolidinyl)-1-deoxy-D-fructose, 1-Deoxy-1-L-proline-D-fructose, 1-Deoxy-1-L-prolino-D-fructose;
SMIL C1C[C@H](N(C1)CC(=O)[C@H]([C@@H]([C@@H](CO)O)O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Fructose-proline (mixture of diastereomers)
Renhedsgrad noter HPLC
InChI formel InChI=1S/C11H19NO7/c13-5-8(15)10(17)9(16)7(14)4-12-3-1-2-6(12)11(18)19/h6,8-10,13,15-17H,1-5H2,(H,18,19)/t6-,8+,9+,10+/m0/s1
Mængde 100 mg
Formel vægt 277.1162
Procent renhed >95%