Få mere at vide
Fructose-leucine (Mixture of Diastereomers), TRC
Artikelnummer. 17957144
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17957144 25 mg -
17948214 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17957144 Leverandør Toronto Research Chemicals Leverandørnr. TRCF79254025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 34393-18-5
Molekylær formel C12H23NO7
Molekylvægt (g/mol) 293.31
Synonym N-(1-Deoxy-D-fructos-1-yl)-L-leucine, (S)-1-[(1-Carboxy-3-methylbutyl)amino]-1-deoxy-D-fructose, D-1-[(L-1-Carboxy-3-methylbutyl)amino]-1-deoxy-fructose, 1-Deoxy-1-L-leucino-D-fructose, Fructoseleucine;
SMIL CC(C)C[C@H](C(=O)O)NCC1([C@H]([C@@H]([C@@H](CO1)O)O)O)O
Tekniske data
Kemisk navn eller materiale Fructose-leucine (Mixture of Diastereomers)
Renhedsgrad noter HPLC
InChI formel InChI=1S/C12H23NO7/c1-6(2)3-7(11(17)18)13-5-12(19)10(16)9(15)8(14)4-20-12/h6-10,13-16,19H,3-5H2,1-2H3,(H,17,18)/t7-,8-,9-,10+,12?/m1/s1
Mængde 25 mg
Formel vægt 293.1475
Procent renhed >95%