Få mere at vide
Fructosazine, TRC
Artikelnummer. 30012509
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012509 25 mg
30010190 10 mg
30004403 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012509 Leverandør Toronto Research Chemicals Leverandørnr. TRCF78500025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13185-73-4
Molekylær formel C12 H20 N2 O8
Molekylvægt (g/mol) 320.3
Synonym (1R,1'R,2S,2'S,3R,3'R)-1,1'-(2,5-Pyrazinediyl)bis-1,2,3,4-butanetetrol;
IUPAC navn (1R,2S,3R)-1-[5-[(1R,2S,3R)-1,2,3,4-tetrahydroxybutyl]pyrazin-2-yl]butane-1,2,3,4-tetrol
SMIL OC[C@@H](O)[C@@H](O)[C@H](O)c1cnc(cn1)[C@@H](O)[C@H](O)[C@H](O)CO
Tekniske data
Kemisk navn eller materiale Fructosazine
Renhedsgrad noter HPLC
Mængde 25 mg
InChI formel InChI=1S/C12H20N2O8/c15-3-7(17)11(21)9(19)5-1-13-6(2-14-5)10(20)12(22)8(18)4-16/h1-2,7-12,15-22H,3-4H2/t7-,8-,9-,10-,11-,12-/m1/s1
Formel vægt 320.122
Procent renhed >95%