Learn More
Memmert™ Fresh-Air Filter for UF55
Fresh-air filter designed to be mounted on the bottom of the unit. Castor frame or subframe not included.
Brand: Memmert™ UF55R8
Additional Details : Weight : 0.01200kg
Description
- Requires castor frame or subframe
- Filtration efficiency 80%
- Mounted at the bottom of the appliance
Specifications
UF55 Oven | |
Fresh Air Filter |