Learn More
Memmert™ Fresh-Air Filter for UF450plus
Fresh-air filter designed to be mounted on the bottom of the unit. Castor frame or subframe not included.
Brand:  Memmert™ UF450plusR8
Additional Details : Weight : 0.01200kg
Product Code. 17539415
  • 2970.00 DKK / Each
Estimated Shipment: 01-07-2024
to see stock.
Add to basket
Description
Description
  • Filtration efficiency 80%
  • Mounted at the bottom of the appliance
Specifications
Specifications
UF450plus Oven
Fresh Air Filter