missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXB2 Antibody (PCRP-FOXB2-2B2) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14125
Additional Details : Weight : 0.00970kg
Description
FOXB2 Monoclonal antibody specifically detects FOXB2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein ArraySpecifications
FOXB2 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1 to 2 μg /million cells, ELISA, Immunoprecipitation 1 to 2 μg per 100-500 μg protein, Protein Array | |
bA159H20.4, forkhead box B2, forkhead box protein B2 | |
Recombinant fragment (around aa1-200) of human FOXB2 protein (exact sequence is proprietary) (Uniprot: Q5VYV0) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunoprecipitation, Peptide Array | |
PCRP-FOXB2-2B2 | |
Unconjugated | |
10mM PBS without Preservative | |
Mouse | |
Protein A or G purified | |
RUO | |
442425 | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IgG2a |