Få mere at vide
Cole Parmer Fowler Micrometer Calibration Set
Artikelnummer. 11789789 Shop alle Cole-Parmer produkter
Skift visning
Pakningsstørrelse:
Hver
Artikelnummer. Pakningsstørrelse
11789789 Hver
1 options
Artikelnummer. 11789789 Leverandør Cole Parmer Leverandørnr. 9723602
Sikkerhed starter her: Spar op til 60 %
Brug rabatkode 28008
Se alle kampagner
Forventet forsendelse: 28-01-2027
Fowler Micrometer Calibration Set is ideal for maintenance personnel and technicians, as well as beginner and professional users.
  • A ring gauge is used to measure, verify and/or calibrate gauge diameter sizes.
  • Ring gauge kits are offered in sizes ranging from 0.0625 in. up to 4 in. and are available with standard steel blocks or for abrasion and corrosion resistance choose zirconium-based ceramic blocks.
  • For the most accurate measurements, NIST-traceable calibration services for your gauge blocks are provided.
  • Calibration Gauge Block Kits are available with steel blocks or zirconium-based ceramic blocks that are abrasion- and corrosion-resistant.
  • Kits include a protective wooden case, and NIST traceable Certificate
Tekniske data
Produkttype Lineal