missing translation for 'onlineSavingsMsg'
Learn More

Duran™ Baffled Flask

Provide increased agitation at lower speeds for a more gentle medium aeration of sensitive cell cultures. Duran™ Four Bottom Borosilicate Glass Baffled Flask molded bottom increases aeration for better growth, especially with bacteria.

Brand:  Duran™ 212834405

Additional Details : Weight : 0.02400kg

 View more versions of this product

Product Code. 15854498

  • 450.00 DKK / Pack of 4


Estimated Shipment
07-06-2019
Add to basket

Description

Description

  • Consistent flask shapes for highly reproducible experimental conditions and results
  • Borosillicate glass with high chemical and thermal shock resistance
  • GL 45 thread necks
  • Ehrlenmeyer
  • Four bottom baffles increase aeration at higher shaking speeds
Specifications

Specifications

USP Standard
105mm
Borosilicate Glass
GL45 Thread
Screw cap
500 mL
180mm
Videos