Få mere at vide
Fosnetupitant, TRC
Artikelnummer. 30412570
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412570 50 mg
30420671 100 mg
30419521 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30412570 Leverandør Toronto Research Chemicals Leverandørnr. TRCP83173050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1703748-89-3
Molekylær formel C31H35F6N4O5P
Molekylvægt (g/mol) 688.6
Synonym 07PNET, {4-[5-{2-[3,5-bis(trifluoromethyl)phenyl]-N,2-dimethylpropanamido}-4-(2-methylphenyl)pyridin-2-yl]-1-methylpiperazin-1-ium-1-yl}methyl hydrogen phosphate
IUPAC navn [4-[5-[[2-[3,5-bis(trifluoromethyl)phenyl]-2-methylpropanoyl]-methylamino]-4-(2-methylphenyl)pyridin-2-yl]-1-methylpiperazin-1-ium-1-yl]methyl hydrogen phosphate
SMIL CN(C(=O)C(C)(C)c1cc(cc(c1)C(F)(F)F)C(F)(F)F)c2cnc(cc2c3ccccc3C)N4CC[N+](C)(COP(=O)(O)[O-])CC4
Tekniske data
Kemisk navn eller materiale Fosnetupitant
Mængde 50 mg
InChI formel InChI=1S/C31H35F6N4O5P/c1-20-8-6-7-9-24(20)25-17-27(40-10-12-41(5,13-11-40)19-46-47(43,44)45)38-18-26(25)39(4)28(42)29(2,3)21-14-22(30(32,33)34)16-23(15-21)31(35,36)37/h6-9,14-18H,10-13,19H2,1-5H3,(H-,43,44,45)
Formel vægt 688.2249