Få mere at vide
Fosinopril USP Related Compound H, Mikromol™
Artikelnummer. 30051598
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30051598 25 mg
30050830 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30051598 Leverandør Mikromol Leverandørnr. MM0645.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Fosinopril USP Related Compound H
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-phenylbutyl phosphonic acid
CAS 46348-61-2
Analyt- eller komponentnavne 4-phenylbutyl phosphonic acid
Forsendelsestilstand Room Temperature
Molekylær formel C10 H15 O3 P
Certificeringer/compliance ISO 17025
Synonym (4-Phenylbutyl)phosphonic Acid, P-(4-Phenylbutyl)phosphonic Acid, Phosphonic acid, (4-phenylbutyl)- (9CI), 4-Phenylbutylphosphonic acid, Fosinopril Related Compound H
InChI formel InChI=1S/C10H15O3P/c11-14(12,13)9-5-4-8-10-6-2-1-3-7-10/h1-3,6-7H,4-5,8-9H2,(H2,11,12,13)
SMIL OP(=O)(O)CCCCc1ccccc1
IUPAC navn 4-phenylbutylphosphonic acid
Molekylvægt (g/mol) 214.2
Mængde 25 mg
Formel vægt 214.0759 g/mol