missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, Puriss., meets analytical spec.s of DAC, FCC, 98.0-100%, Honeywell Fluka™

Brand:  Honeywell Fluka 27001-6X500ML

Additional Details : CAS Number : 64-18-6 Weight : 3.60000kg Transport : UN number : 1779 Chem class : 1a Pack group : II

Quantity 6 × 500mL

 View more versions of this product

Product Code. 15642830

  • 960.86 DKK / Pack of 6


Estimated Shipment date
from Supplier 27-02-2018
Add to basket

Specifications

Chemical Identifiers

64-18-6
1.22g/mL at 25°C
46.03
0.57
1004°F
MFCD00003297
UN1779
44.8mmHg (20°C)
Formic Acid
98.0 to 100%
puriss.
48°C (118.4°F)
HCOOH
Meets analytical specifications of DAC, FCC
1.6 (vs air)

Specifications

Formic Acid
64-18-6
98.0 to 100%
46.03
UN1779
0.57
44.8mmHg (20°C)
6 × 500mL
Meets analytical specifications of DAC, FCC
puriss.
HCOOH
MFCD00003297
1.22g/mL at 25°C
1004°F
1.6 (vs air)
48°C (118.4°F)
Videos
SDS