missing translation for 'onlineSavingsMsg'
Learn More
Formic Acid, Pure, about 85%, Fisher Chemical™ Product Code.: 10114433

Formic Acid, Pure, about 85%, Fisher Chemical™

Product Code. p-7073030 Shop All Fisher Chemical Products
1 L, HDPE plastic bottle, 1 L
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.50 L
25 L
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 10114433

Brand: Fisher Chemical F/1750/PB15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Formic Acid, Pure, about 85%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-18-6
Molecular Formula CH2O2
Molecular Weight (g/mol) 46.025
MDL Number 3297
InChI Key BDAGIHXWWSANSR-UHFFFAOYSA-N
Synonym methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglattShow MoreShow Less
PubChem CID 284
ChEBI CHEBI:30751
IUPAC Name formic acid
SMILES C(=O)O

Specifications

pH 2.1
Melting Point 8°C
Boiling Point 101°C
Color Colorless
Formula Weight 46.02g/mol
Quantity 1 L
Packaging HDPE plastic bottle
Physical Form Liquid
Vapor Pressure 44mbar at 20°C

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Product Code
Lot Number2537183 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeF/1750/PB17,F/1750/25,F/1750/PB15
Lot Number2536450 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeF/1750/PB17,F/1750/25,F/1750/PB15
Lot Number2529885 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeF/1750/PB17,F/1750/25,F/1750/PB15
Lot Number2528527 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeF/1750/PB17,F/1750/25,F/1750/PB15
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formic acid about 85%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Formic Acid, Pure, about 85%, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.