Få mere at vide
Formamide-1-d1, CDN
Artikelnummer. 30320532
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30320532 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320532 Leverandør CDN Leverandørnr. CDND03401G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Formamide-1-D
CAS 35692-88-7
Molekylær formel C D H2 N O
Mængde 1 g
Synonym Formamide-1-d (7 CI, 9 CI), Form-d-amide (6 CI), Formamide-d1, Formamide-1-D, [1-2 H]Formamide, Carbamaldehyde-d, Form-d-imidic acid, Formamide-1-d1
InChI formel InChI=1 S/CH3NO/c2-1-3/h1H,(H2,2,3)/i1D
SMIL [2 H]C(=O)N
IUPAC navn deuterioformamide
Molekylvægt (g/mol) 46.0468
Formel vægt 46.0277 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity