Få mere at vide
Formaldehyde-13C,D2 (20% in D2O), TRC
Artikelnummer. 30407583
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30407583 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407583 Leverandør Toronto Research Chemicals Leverandørnr. TRCF691353100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Formaldehyde-13C,D2
CAS 63101-50-8
Molekylær formel 13C 2H2 O
Mængde 100 mg
Synonym Formaldehyde-13C-d2 (9CI), Formaldehyde-d2-13C, Formaldehyde-13C,D2, Formaldehyde-13C,d2, Methanal-13C,d2
InChI formel InChI=1S/CH2O/c1-2/h1H2/i1+1D2
SMIL [2H][13C](=O)[2H]
IUPAC navn dideuterio(1^{13}C)methanone
Molekylvægt (g/mol) 33.03
Formel vægt 33.0265
Koncentration eller sammensætning (efter analyt eller komponenter) 20 %