Learn More
BRAND™ Forceps
PTFE forceps
Brand: BRAND™ 113125
296.96 DKK valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Specifications
Rectangular | |
PTFE | |
100 mm | |
1 |