Få mere at vide
Folinic Acid, TRC
Artikelnummer. 30137272
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137272 1 mg
30008829 5 mg
30011114 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137272 Leverandør Toronto Research Chemicals Leverandørnr. TRCF6803401MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 58-05-9
Molekylær formel C20H23N7O7
Molekylvægt (g/mol) 473.45
Synonym 5-Formyl-5,6,7,8-tetrahydrofolic Acid, 5-Formyltetrahydrofolic Acid, 5-Formyltetrahydropteroylglutamic Acid, CF, Folinic Acid, Folinic acid-SF, Kunyrin, Leucal, Leucoverin, Leucovorin, N5-Formyl-5,6,7,8-tetrahydrofolic Acid, N5-Formyl-THF, N5-Formyltetrahydrofolic Acid, Welcovorin, l-Leucovorin, (2S)-2-(4-(((2-Amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl)amino)benzamido)pentanedioic Acid
SMIL NC1=NC(=O)C2=C(NCC(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)O)C(=O)O)N2C=O)N1
Tekniske data
Kemisk navn eller materiale Folinic Acid
Renhedsgrad noter HPLC
InChI formel InChI=1S/C20H23N7O7/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32)/t12?,13-/m0/s1
Mængde 1 mg
Procent renhed >95%