Få mere at vide
Uvex Fold-flat Mask silv-Air Premium 5310+ FFP3
Shop alle Uvex produkter
:
Filter Type: FFP3
Beskrivelse
General Features
- Individually packed
- Color: white
- With exhalation valve
Safety Features
- Certified FFP3 face mask to EN 149
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Cannot be reused
Comfort Features
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
- Soft sealing lip in the nose area guarantees a secure, comfortable sealed position of the mask
- Adjustable head band offering easy handling and comfortable fit
- Flexible and adjustable nose clip
- Soft material edges for high wearing comfort
Tekniske data
Tekniske data
| Certificeringer/overholdelse (Europa) | EN 149 (D marked) |
| Engangs | Single-use |
| Materiale | Polypropylene, Polyester |
| Med udåndingsventil | Yes |
| Filtereffektivitet | FFP3 |
| Sterilitet | Non-sterile |