Få mere at vide
Uvex Fold-flat Mask silv-Air Premium 5210+ FFP2
Artikelnummer. 17562975 Shop alle Uvex produkter
Skift visning
Filter Type: FFP2
Artikelnummer.
17562975 Filter Type: FFP2
1 options
Artikelnummer. 17562975 Leverandør Uvex Leverandørnr. 8765211
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
The silv-Air premium+ product line comprises classic folding respirators that adapt to different face shapes which fits perfectly to larger face shapes. The headband and integrated sealing lip in all protection classes mean the masks are easy to put on correctly as well as making them comfortable to wear, even for extended periods.
General Features
  • Individually packed
  • Color: white
  • With exhalation valve
Safety Features
  • Certified FFP2 face mask to EN 149:2001+A1:2009
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Cannot be reused
Comfort Features
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
  • Soft sealing lip in the nose area guarantees a secure, comfortable sealed position of the mask
  • Adjustable head band offering easy handling and comfortable fit
  • Flexible and adjustable nose clip
  • Soft material edges for high wearing comfort
TRUSTED_SUSTAINABILITY
Tekniske data
Certificeringer/overholdelse (Europa) EN 149 (D marked)
Engangs Single-use
Materiale Polypropylene, Polyester
Med udåndingsventil Yes
Filtereffektivitet FFP2
Sterilitet Non-sterile