Få mere at vide
Uvex Fold-flat Mask silv-Air c 3200 FFP2
Shop alle Uvex produkter
:
Filter Type: FFP2
Beskrivelse
General Features
- Particle-filtering folding mask
- Color: white
Protection Features
- Certified FFP2 face mask to EN 149:2001+A1:2009
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Cannot be reused
Comfort Features
- Seamless headband and flexible, adjustable nose clip
- Comfortable sealing lip in nose area for secure fit
Tekniske data
Tekniske data
| Certificeringer/overholdelse (Europa) | EN 149 (D marked) |
| Design Type | Flat-fold |
| Engangs | Single-use |
| Materiale | Polypropylene, Polyester |
| Størrelse | Standard |
| Med udåndingsventil | No |
| Filtereffektivitet | FFP2 |
| Sterilitet | Non-sterile |