missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress FMRF

Product Code. 16451948
Encompass_Preferred
Change view
Click to view available options
Mængde:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16451948 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451948 Supplier MedChemExpress Supplier No. HYP0293

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress FMRF is a peptide consisting of 4 amino acid residues.

Specifications

Renhed 99.08%
CAS 74012-06-9
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molekylvægt (g/mol) 599.74 g/mol
Produkttype Peptides
Mængde 5 mg
Forsendelsestilstand Room temperature
Opløselighed DMSO : 100 mg/mL (166.74 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.