Få mere at vide
Fmoc-L-homocitrulline, TRC
Artikelnummer. 30403588
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30403588 250 mg
30396285 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30403588 Leverandør Toronto Research Chemicals Leverandørnr. TRCF679730250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fmoc-L-homocitrulline
CAS 201485-17-8
Molekylær formel C22H25N3O5
Mængde 250 mg
InChI formel InChI=1S/C22H25N3O5/c23-21(28)24-12-6-5-11-19(20(26)27)25-22(29)30-13-18-16-9-3-1-7-14(16)15-8-2-4-10-17(15)18/h1-4,7-10,18-19H,5-6,11-13H2,(H,25,29)(H,26,27)(H3,23,24,28)/t19-/m0/s1
SMIL C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)N[C@@H](CCCCNC(=O)N)C(=O)O
Molekylvægt (g/mol) 411.45