Få mere at vide
Fmoc-L-cyclopropylalanine, TRC
Artikelnummer. 30179340
Skift visning
Mængde:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179340 250 mg
30179076 1 g
30179134 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30179340 Leverandør Toronto Research Chemicals Leverandørnr. TRCF622780250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 214750-76-2
Molekylær formel C21 H21 N O4
Molekylvægt (g/mol) 351.4
Synonym Fmoc-β-cyclopropyl-L-Alanine, N-Fmoc-cyclopropyl alanine, N-(9-fluorenylmethoxycarbonyl)-3-cyclopropyl-L-alanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-cyclopropylpropanoic Acid
IUPAC navn (2S)-3-cyclopropyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMIL OC(=O)[C@H](CC1CC1)NC(=O)OCC2c3ccccc3c4ccccc24
Tekniske data
Kemisk navn eller materiale Fmoc-L-cyclopropylalanine
InChI formel InChI=1S/C21H21NO4/c23-20(24)19(11-13-9-10-13)22-21(25)26-12-18-16-7-3-1-5-14(16)15-6-2-4-8-17(15)18/h1-8,13,18-19H,9-12H2,(H,22,25)(H,23,24)/t19-/m0/s1
Mængde 250 mg
Formel vægt 351.1471