missing translation for 'onlineSavingsMsg'
Learn More

Fmoc-Aph(Hor)-OH (~10% THF) (Degarelix intermediate), TRC

Product Code. 30179488
Change view
Click to view available options
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30179488 500 mg
30179000 100 mg
30179058 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179488 Supplier Toronto Research Chemicals Supplier No. TRCF647563500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1253282-31-3
Molekylær formel C29 H26 N4 O7
Molekylvægt (g/mol) 542.54
Synonym N-[(9H-fluoren-9-ylmethoxy)carbonyl]-4-[[[(4S)-hexahydro-2,6-dioxo-4-pyrimidinyl]carbonyl]amino]-L-Phenylalanine
IUPAC navn (2S)-3-[4-[[(4S)-2,6-dioxohexahydropyrimidine-4-carbonyl]amino]phenyl]-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMIL OC(=O)[C@H](Cc1ccc(NC(=O)[C@@H]2CC(=O)NC(=O)N2)cc1)NC(=O)OCC3c4ccccc4c5ccccc35

Specifications

Kemisk navn eller materiale Fmoc-Aph(Hor)-OH
InChI formel InChI=1S/C29H26N4O7/c34-25-14-23(31-28(38)33-25)26(35)30-17-11-9-16(10-12-17)13-24(27(36)37)32-29(39)40-15-22-20-7-3-1-5-18(20)19-6-2-4-8-21(19)22/h1-12,22-24H,13-15H2,(H,30,35)(H,32,39)(H,36,37)(H2,31,33,34,38)/t23-,24-/m0/s1
Mængde 500 mg
Formel vægt 542.1801
Forsendelsestilstand Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.