Få mere at vide
Tocris Bioscience™ FMK 9a
Beskrivelse
FMK 9a is a potent ATG4B/autophagin-1 inhibitor (IC50 values are 73 nM and 80 nM in cell-based LRA and TR-FRET assays, respectively). Also inhibits calpain and cathepsin B (IC50 values are 96 nM and 200 nM, respectively).
Tekniske data
Tekniske data
| Kemisk navn eller materiale | N-[(1S)-2-[(3-Fluoro-2-oxopropyl)amino]-2-oxo-1-(phenylmethyl)ethyl]-1-naphthalenecarboxamide |
| CAS | 1955550-51-2 |
| Mængde | 2 mg |
| Mål | Additional Synthetic Protease Inhibitors |
| Molekylær formel | C23H21FN2O3 |
| Renhed | 90% |