Få mere at vide
Fluvastatin Methyl Ester, Mikromol™
Beskrivelse
Fluvastatin Methyl Ester
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Fluvastatin Methyl Ester |
| CAS | 93957-53-0 |
| Analyt- eller komponentnavne | Fluvastatin Methyl Ester |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C25 H28 F N O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Fluvastatin Methyl Ester, (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic acid methyl ester, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, methyl ester, [R*,S*-(E)]-(+/-)-, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, methyl ester, [R*,S*-(E)]- |
| InChI formel | InChI=1S/C25H28FNO4/c1-16(2)27-22-7-5-4-6-21(22)25(17-8-10-18(26)11-9-17)23(27)13-12-19(28)14-20(29)15-24(30)31-3/h4-13,16,19-20,28-29H,14-15H2,1-3H3/b13-12+/t19-,20-/m1/s1 |
| SMIL | COC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(c2ccc(F)cc2)c3ccccc3n1C(C)C |
| IUPAC navn | methyl (E,3R,5S)-7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-3,5-dihydroxyhept-6-enoate |
| Vis mere |