Få mere at vide
Fluvastatin EP Impurity B (as Racemate), Mikromol™
Beskrivelse
Fluvastatin EP Impurity B (as Racemate)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 1,1-Dimethylethyl (3RS,5SR,6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate (Fluvastatin tert-Butyl Ester) |
| CAS | 129332-29-2 |
| Analyt- eller komponentnavne | 1,1-Dimethylethyl (3RS,5SR,6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate (Fluvastatin tert-Butyl Ester) |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C28 H34 F N O4 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Fluvastatin Sodium Imp. B (EP), Fluvastatin Imp. B (EP), Fluvastatin USP Related Compound B, Fluvastatin USP RC B, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, 1,1-dimethylethyl ester, (3R,5S,6E)-rel-, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, 1,1-dimethylethyl ester, [R*,S*-(E)]-(+/-)-, 6-Heptenoic acid, 7-[3-(4-fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxy-, 1,1-dimethylethyl ester, [R*,S*-(E)]-, tert-Butyl Fluvastatin, 1,1-Dimethylethyl (3RS,5SR,6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate, Fluvastatin tert-Butyl Ester, tert-Butyl (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-isopropyl-1H-indol-2-yl]-3,5-dihydroxyhept-6-enoate, Fluvastatin Related Compound B, Fluvastatin Sodium Impurity B as Racemate, Fluvastatin Impurity B as Racemate |
| InChI formel | InChI=1S/C28H34FNO4/c1-18(2)30-24-9-7-6-8-23(24)27(19-10-12-20(29)13-11-19)25(30)15-14-21(31)16-22(32)17-26(33)34-28(3,4)5/h6-15,18,21-22,31-32H,16-17H2,1-5H3/b15-14+/t21-,22-/m1/s1 |
| SMIL | CC(C)n1c(\C=C\[C@@H](O)C[C@@H](O)CC(=O)OC(C)(C)C)c(c2ccc(F)cc2)c3ccccc13 |
| IUPAC navn | tert-butyl (E,3R,5S)-7-[3-(4-fluorophenyl)-1-propan-2-ylindol-2-yl]-3,5-dihydroxyhept-6-enoate |
| Vis mere |