Få mere at vide
Fluvastatin-d7 Sodium Salt, TRC
Artikelnummer. 30404068
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30404068 1 mg
30395875 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404068 Leverandør Toronto Research Chemicals Leverandørnr. TRCF6012541MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fluvastatin-d7 Sodium Salt
CAS 1260178-87-7
Molekylær formel C24H18D7FNNaO4
Mængde 1 mg
Synonym (3R,5S,6E)-rel-7-[3-(4-Fluorophenyl)-1-(1-methylethyl-d7)-1H-indol-2-yl]-3,5-dihydroxy-6-heptenoic Acid Sodium Salt, Fluindostatin-d7, XU 62-320-d7, Lescol-d7, Lipaxan-d7, Primexin-d7
InChI formel InChI=1S/C24H26FNO4.Na/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30;/h3-12,15,18-19,27-28H,13-14H2,1-2H3,(H,29,30);/q;+1/p-1/b12-11+;/t18-,19-;/m1./s1/i1D3,2D3,3D,6D,15D;
SMIL FC(C=C1)=CC=C1C2=C(/C=C/[C@@H](O)C[C@@H](O)CC([O-])=O)N(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C3=C2C=CC=C3.[Na+]
Molekylvægt (g/mol) 440.49