Få mere at vide
Fluroxypyr-1-methylheptyl Ester, TRC
Artikelnummer. 30012562
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012562 5 g
17961515 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30012562 Leverandør Toronto Research Chemicals Leverandørnr. TRCF5987905G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81406-37-3
Molekylær formel C15 H21 Cl2 F N2 O3
Molekylvægt (g/mol) 367.24
Synonym Acetic acid, [(4-amino-3,5-dichloro-6-fluoro-2-pyridinyl)oxy]-, 1-methylheptyl ester (9CI), Attain A, Dowco 433, Fluroxypyr 1-methylheptyl ester, Fluroxypyr meptyl, Fluroxypyr meptyl ester, Spotlight, Spotlight (pyridinoxyacetate herbicide), Starane, Starane 2, Starane 250, Starane 250EC, Starane Ultra, Tomahawk, Trophy A, XRM 5084, Acetic acid, 2-[(4-amino-3,5-dichloro-6-fluoro-2-pyridinyl)oxy]-, 1-methylheptyl ester, Fluroxypyr-1-methylheptyl Ester
IUPAC navn octan-2-yl 2-(4-amino-3,5-dichloro-6-fluoropyridin-2-yl)oxyacetate
SMIL CCCCCCC(C)OC(=O)COc1nc(F)c(Cl)c(N)c1Cl
Tekniske data
Kemisk navn eller materiale Fluroxypyr-1-methylheptylester
Mængde 5 g
InChI formel InChI=1S/C15H21Cl2FN2O3/c1-3-4-5-6-7-9(2)23-10(21)8-22-15-12(17)13(19)11(16)14(18)20-15/h9H,3-8H2,1-2H3,(H2,19,20)
Formel vægt 366.0913