missing translation for 'onlineSavingsMsg'
Få mere at vide

Fluridone, TRC

Artikelnummer. 30400933
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30400933 250 mg
30396328 100 mg
30399970 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400933 Supplier Toronto Research Chemicals Supplier No. TRCF598770250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59756-60-4
Molekylær formel C19 H14 F3 N O
Molekylvægt (g/mol) 329.32
Synonym Fluridone, 4(1H)-Pyridinone, 1-methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]- (9CI, ACI), 1-Methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]-4(1H)-pyridinone (ACI), EL 171, Fluridone, Sonar
IUPAC navn 1-methyl-3-phenyl-5-[3-(trifluoromethyl)phenyl]pyridin-4-one
SMIL CN1C=C(C(=O)C(=C1)c2cccc(c2)C(F)(F)F)c3ccccc3

Specifications

Kemisk navn eller materiale Fluridone
Mængde 250 mg
InChI formel InChI=1S/C19H14F3NO/c1-23-11-16(13-6-3-2-4-7-13)18(24)17(12-23)14-8-5-9-15(10-14)19(20,21)22/h2-12H,1H3
Formel vægt 329.1027
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.