Få mere at vide
(±)-Flurbiprofen-d5 (phenyl-d5), CDN
Artikelnummer. 30321417
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321417 0.05 g
30319491 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321417 Leverandør CDN Leverandørnr. CDND74050.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Flurbiprofen-d5
CAS 215175-76-1
Molekylær formel C15 D5 H8 F O2
Mængde 0.05 g
Synonym [1,1'-Biphenyl-2',3',4',5',6'-d5]-4-acetic acid, 2-fluoro-α-methyl- (9 CI), 2-Fluoro-α-methyl-[1,1'-biphenyl-2',3',4',5',6'-d5]-4-acetic acid, Flurbiprofen-d5, Flurbiprofen-D5 (phenyl-D5), (±)-Flurbiprofen-d5 (phenyl-d5), 2-(2-Fluorobiphenyl-4-yl-2',3',4',5',6'-d5)propionic acid
InChI formel InChI=1 S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10 H,1H3,(H,17,18)/i2D,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])c2ccc(cc2F)C(C)C(=O)O
IUPAC navn 2-[3-fluoro-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]propanoic acid
Molekylvægt (g/mol) 249.292
Formel vægt 249.121 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity