missing translation for 'onlineSavingsMsg'
Få mere at vide

Fluoromethylcholine Chloride (>90%), TRC

Artikelnummer. 30392700
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30392700 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30392700 Leverandør Toronto Research Chemicals Leverandørnr. TRCF598258100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Fluoromethylcholine Chloride
CAS 459424-38-5
Molekylær formel C5H13ClFNO
Mængde 100 mg
Synonym Fluorocholine Chloride
InChI formel InChI=1S/C5H13FNO.ClH/c1-7(2,5-6)3-4-8;/h8H,3-5H2,1-2H3;1H/q+1;/p-1
SMIL [Cl-].C[N+](C)(CF)CCO
Molekylvægt (g/mol) 157.61
Formel vægt 157.067
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.