Learn More
Jenway™ Fluorimeter Interference Filter
Requires a much more narrow range of wavelengths. Jenway™ Fluorimeter Interference Filter allows a specified wavelength and light to pass through to the sample.
Brand: Jenway™ 10091-24
Additional Details : Weight : 0.10000kg
Description
- For use with Jenway™ Fluorimeters
- Normally used as the emission filters where a much narrower range of wavelengths is required
- Allow the specified wavelength and light ±5nm of this through to the sample.
Specifications
Interference Filter | |
320 nm | |
Jenway Fluorimeters |