Få mere at vide
Fluoride Standard: F- @ 10000 μg/mL in H2O, VHG™
Artikelnummer. 17978474
Skift visning
Mængde:
500 ml
Artikelnummer. Mængde
17978474 500 ml
1 options
Artikelnummer. 17978474 Leverandør VHG Labs Leverandørnr. VHGI1PF500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Fluoride Standard: F- @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fluoride
CAS 16984-48-8
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Fluoride
Forsendelsestilstand Room Temperature
Molekylær formel F-
Certificeringer/compliance ISO 17034
Synonym F−
SMIL [F-]
IUPAC navn Fluoride
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 19
Mængde 500 ml
Formel vægt 19 g/mol