Få mere at vide
Fluoride Ion Chromatography Standard: F- @ 100μg/mL in H2O, VHG™
Artikelnummer. 17949914
Skift visning
Mængde:
100 ml
500 ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17949914 100 ml
17978874 500 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17949914 Leverandør VHG Labs Leverandørnr. VHGIF100100
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Fluoride Ion Chromatography Standard: F- @ 100μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fluoride
CAS 16984-48-8
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Fluoride
Forsendelsestilstand Room Temperature
Molekylær formel F-
Certificeringer/compliance ISO 17034
Synonym F−
SMIL [F-]
IUPAC navn Fluoride
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 19
Mængde 100 ml
Formel vægt 19 g/mol