missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein/Oregon Green Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen A889
Additional Details : Weight : 0.10000kg
Description
Fluorescein/Oregon Green Polyclonal antibody specifically detects Fluorescein/Oregon Green in Chemical samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry.Specifications
Fluorescein/Oregon Green | |
Polyclonal | |
Unconjugated | |
FITC | |
Fluorescein/Oregon Green | |
500 μL | |
Primary | |
Antibody | |
IgG |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
1 mg/mL | |
0.1M potassium phosphate with 5mM sodium azide; pH 8 | |
Rabbit | |
purified | |
RUO | |
4° C | |
Liquid | |
Chemical |