Få mere at vide
MedChemExpress Fluorescein di(β-D-galactopyranoside)
Tekniske data
Tekniske data
| Mængde | 1 mg |
| Produkttype | Dye Reagents |
| Molekylvægt (g/mol) | 656.59 g/mol |
| Indhold og opbevaring | -20°C, protect from light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Forsendelsestilstand | Room temperature |
| CAS | 17817-20-8 |
| Synonym | FDG |