missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Fluorescein Antibody, Texas Red, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB110-3648-100UG
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Fluorescein Polyclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay.
Tekniske data
| Fluorescein | |
| Polyclonal | |
| Western Blot >1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence >1:20000, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay >1:20000 | |
| FITC, fluorescein isothyocianate | |
| Fluorescein conjugated to Goat IgG. | |
| 100 μg | |
| Non-species specific | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Texas Red | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free | |
| Goat | |
| Affinity Purified | |
| Fluorescein | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion