Få mere at vide
Fluorene-d10, CDN
Artikelnummer. 30319379
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319379 1 g -
30320904 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319379 Leverandør CDN Leverandørnr. CDND12981G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Fluorene D10
CAS 81103-79-9
Molekylær formel C13 D10
Mængde 1 g
Synonym 9 H-Fluorene-1,2,3,4,5,6,7,8,9,9-d10, Fluorene-d10, Perdeuterofluorene
InChI formel InChI=1 S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8 H,9H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9D2
SMIL [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c([2 H])c([2 H])c3C2([2 H])[2 H]
IUPAC navn 1,2,3,4,5,6,7,8,9,9-decadeuteriofluorene
Molekylvægt (g/mol) 176.2801
Formel vægt 176.141 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity