Få mere at vide
Flunitrazepam EP Impurity B, Mikromol™
Beskrivelse
Flunitrazepam EP Impurity B
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Desmethylflunitrazepam |
| CAS | 2558-30-7 |
| Analyt- eller komponentnavne | Desmethylflunitrazepam |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H10 F N3 O3 |
| Certificeringer/compliance | ISO 17025 |
| Synonym | 5-(2-Fluorophenyl)-7-nitro-1,3-dihydro-2H-1,4-benzodiazepin-2-one (Demethylflunitrazepam), 5-(2-Fluorophenyl)-7-nitro-1,3-dihydro-2H-1,4-benzodiazepin-2-one, Flunitrazepam Imp. B (EP), Demethylflunitrazepam, Desmethylflunitrazepam, Flunitrazepam Impurity B |
| InChI formel | InChI=1S/C15H10FN3O3/c16-12-4-2-1-3-10(12)15-11-7-9(19(21)22)5-6-13(11)18-14(20)8-17-15/h1-7H,8H2,(H,18,20) |
| SMIL | [O-][N+](=O)c1ccc2NC(=O)CN=C(c3ccccc3F)c2c1 |
| IUPAC navn | 5-(2-fluorophenyl)-7-nitro-1,3-dihydro-1,4-benzodiazepin-2-one |
| Vis mere |